1-Ethyl-1-methyl-4-oxopiperidin-1-ium iodide 95%

Supplier: Ambeed
Warning

A171540-1G A171540-250MG
76985-442EA 92.3 USD
76985-442 76985-444
1-Ethyl-1-methyl-4-oxopiperidin-1-ium iodide 95%
1-Ethyl-1-methyl-4-oxopiperidin-1-ium iodide

Precaución: for Research Use Only

Formula: C₈H₁₆INO
MW: 269.12 g/mol
MDL Number: MFCD16620306
CAS Number: 77542-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR