6-Fluoro-2-phenylimidazo[1,2-a]pyridine 97%
Supplier: Ambeed
|
Warning
|
A171448-100MG
A171448-1G
A171448-250MG
A171448-5G
76985-434EA
9.55
USD
76985-434
76985-436
76985-438
76985-440
6-Fluoro-2-phenylimidazo[1,2-a]pyridine 97%
6-Fluoro-2-phenylimidazo[1,2-a]pyridine
Precaución: for Research Use Only
Formula:
C₁₃H₉FN₂ MW: 212.22 g/mol |
MDL Number:
MFCD11845358 CAS Number: 1126635-20-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...