Boc-Ser(tBu)-OH 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-Ser(tBu)-OH, N-Boc-O-tert-butyl-L-serine
A171316-100G
A171316-10G
A171316-1G
A171316-25G
A171316-5G
76998-208EA
95
USD
76998-208
76998-210
76998-212
76998-214
76998-216
Boc-Ser(tBu)-OH 97%
Boc-Ser(tBu)-OH
Precaución: for Research Use Only
Formula:
C₁₂H₂₃NO₅ MW: 261.31 g/mol |
MDL Number:
MFCD00079666 CAS Number: 13734-38-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...