2,3-Diaminobenzoic acid 95%

Supplier: Ambeed
Warning

A171314-1G A171314-25G
77029-032EA 7.43 USD
77029-032 77871-859 77871-860
2,3-Diaminobenzoic acid 95%
2,3-Diaminobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MDL Number: MFCD00137818
CAS Number: 603-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR