(6-Bromo-2-methoxypyridin-3-yl)methanol 98%

Supplier: Ambeed
Warning

A171230-250MG A171230-1G
77011-580EA 96.55 USD
77011-580 77683-392
(6-Bromo-2-methoxypyridin-3-yl)methanol 98%
(6-Bromo-2-methoxypyridin-3-yl)methanol

Precaución: For research use only.

Formula: C₇H₈BrNO₂
MW: 218.04 g/mol
MDL Number: MFCD27922757
CAS Number: 1802489-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR