(S)-2-(2-((tert-Butoxycarbonyl)amino)acetamido)-3-methylbutanoic acid 98%

Supplier: Ambeed
Warning

A171119-1G A171119-250MG
76985-422EA 75 USD
76985-422 76985-424
(S)-2-(2-((tert-Butoxycarbonyl)amino)acetamido)-3-methylbutanoic acid 98%
(S)-2-(2-((tert-Butoxycarbonyl)amino)acetamido)-3-methylbutanoic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₂N₂O₅
MDL Number: MFCD26142782
CAS Number: 28334-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR