6-Bromo-2-chloro-3-nitropyridine 96%

Supplier: Ambeed
Warning

A171080-1G A171080-250MG A171080-5G
77011-570EA 89 USD
77011-570 77011-572 77890-100
6-Bromo-2-chloro-3-nitropyridine 96%
6-Bromo-2-chloro-3-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClN₂O₂
MW: 237.44 g/mol
MDL Number: MFCD27923193
CAS Number: 1430341-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR