Ethyl-5-chloro-1H-pyrrolo[3,2-b]pyridine-2-carboxylate 96%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-chloro-1H-pyrrolo[3,2-b]pyridine-2-carboxylate

A170939-100MG A170939-1G A170939-250MG
77029-026EA 33.95 USD
77029-026 77029-028 77029-030
Ethyl-5-chloro-1H-pyrrolo[3,2-b]pyridine-2-carboxylate 96%
Ethyl-5-chloro-1H-pyrrolo[3,2-b]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂O₂
MW: 224.6437 g/mol
MDL Number: MFCD11109466
CAS Number: 800401-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR