4-{[5-(Trifluoromethyl)-2-pyridinyl]oxy}aniline 95%

Supplier: Ambeed
Danger

Synonyms: 4-((5-(Trifluoromethyl)pyridin-2-yl)oxy)aniline

A170755-10G A170755-1G A170755-5G
76998-194EA 204 USD
76998-194 76998-196 76998-198
4-{[5-(Trifluoromethyl)-2-pyridinyl]oxy}aniline 95%
4-{[5-(Trifluoromethyl)-2-pyridinyl]oxy}aniline

Precaución: for Research Use Only

Formula: C₁₂H₉F₃N₂O
MW: 254.21 g/mol
MDL Number: MFCD03001213
CAS Number: 71422-81-6
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR