7-Boc-2-oxa-7-azaspiro[3.5]nonane 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-oxa-7-azaspiro[3.5]nonane-7-carboxylate, 2-Oxa-7-azaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester

A170657-250MG A170657-100MG
76985-392EA 45 USD
76985-392 77885-341
7-Boc-2-oxa-7-azaspiro[3.5]nonane 97%
7-Boc-2-oxa-7-azaspiro[3.5]nonane

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₃
MW: 227.30 g/mol
MDL Number: MFCD11035887
CAS Number: 240401-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR