2-(2-Methoxy-2-oxoethyl)benzoic acid 98%

Supplier: Ambeed
Warning

A170498-10G A170498-1G A170498-250MG A170498-5G
77029-018EA 680 USD
77029-018 77029-020 77029-022 77029-024
2-(2-Methoxy-2-oxoethyl)benzoic acid 98%
2-(2-Methoxy-2-oxoethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MDL Number: MFCD00661540
CAS Number: 14736-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR