tert-Butyl (4-acetylphenyl)carbamate 96%

Supplier: Ambeed
Warning

A170435-100MG A170435-250MG A170435-1G A170435-5G
77029-014EA 9 USD
77029-014 77029-016 77667-384 77871-855
tert-Butyl (4-acetylphenyl)carbamate 96%
tert-Butyl (4-acetylphenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₃
MW: 235.27 g/mol
MDL Number: MFCD09817646
CAS Number: 232597-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR