Benzyl-2,4-difluorophenylcarbamate 97%

Supplier: Ambeed
Warning

Synonyms: Benzyl (2,4-difluorophenyl)carbamate

A170405-1G A170405-250MG
76985-384EA 117 USD
76985-384 76985-386
Benzyl-2,4-difluorophenylcarbamate 97%
Benzyl-2,4-difluorophenylcarbamate

Precaución: for Research Use Only

Formula: C₁₄H₁₁F₂NO₂
MW: 263.25 g/mol
MDL Number: MFCD21648243
CAS Number: 112434-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR