Boc-D-Asp(OtBu)-OH 97%

Supplier: Ambeed
Warning

A170283-100G A170283-1G A170283-250MG A170283-5G A170283-10G A170283-25G
77011-562EA 465.74 USD
77011-562 77011-564 77011-566 77011-568 77593-054 77611-658
Boc-D-Asp(OtBu)-OH 97%
Boc-D-Asp(OtBu)-OH

Precaución: for Research Use Only

Formula: C₁₃H₂₃NO₆
MW: 289.3 g/mol
MDL Number: MFCD01861313
CAS Number: 155542-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR