N,N-Dibenzylglycine ethyl ester 97%

Supplier: Ambeed
Warning

Synonyms: N,N-Dibenzylglycine Ethyl Ester, Di-Bzl-Gly-OEt, Ethyl 2-(dibenzylamino)acetate

A169975-1G A169975-25G A169975-5G A169975-10G
76998-176EA 6.37 USD
76998-176 76998-178 76998-180 77671-408
N,N-Dibenzylglycine ethyl ester 97%
N,N-Dibenzylglycine ethyl ester

Precaución: for Research Use Only

Formula: C₁₈H₂₁NO₂
MW: 283.37 g/mol
MDL Number: MFCD00796343
CAS Number: 77385-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR