1-(1H-Indazol-5-yl)ethanone 97%

Supplier: Ambeed
Warning

Synonyms: 5-Acetyl-1H-indazole

A169918-250MG A169918-1G
77028-994EA 46.68 USD
77028-994 77667-374
1-(1H-Indazol-5-yl)ethanone 97%
1-(1H-Indazol-5-yl)ethanone

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MW: 160.18 g/mol
MDL Number: MFCD11869763
CAS Number: 1001906-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR