4-Bromo-3-methyl-5-(trifluoromethyl)-1H-pyrazole 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-3-methyl-5-(trifluoromethyl)pyrazole

A169805-10G A169805-1G A169805-25G A169805-5G
76985-358EA 74.27 USD
76985-358 76985-360 76985-362 77611-654
4-Bromo-3-methyl-5-(trifluoromethyl)-1H-pyrazole 97%
4-Bromo-3-methyl-5-(trifluoromethyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₅H₄BrF₃N₂
MW: 229.00 g/mol
MDL Number: MFCD08060951
CAS Number: 60061-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR