3-Chloro-5-methylpyrazin-2(1H)-one 95%

Supplier: Ambeed
Warning

A169802-100MG A169802-1G A169802-250MG
76985-352EA 87 USD
76985-352 76985-354 76985-356
3-Chloro-5-methylpyrazin-2(1H)-one 95%
3-Chloro-5-methylpyrazin-2(1H)-one

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O
MW: 144.56 g/mol
MDL Number: MFCD15144362
CAS Number: 89283-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR