Methyl 3-bromo-5-fluoroisonicotinate 97%

Supplier: Ambeed
Warning

A169525-100MG A169525-1G A169525-250MG
76985-346EA 21 USD
76985-346 76985-348 76985-350
Methyl 3-bromo-5-fluoroisonicotinate 97%
Methyl 3-bromo-5-fluoroisonicotinate

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₂
MDL Number: MFCD14698118
CAS Number: 1214325-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR