9,9-Di-n-dodecyl-2,7-dibromofluorene 97%

Supplier: Ambeed
Warning

Synonyms: 2,7-Dibromo-9,9-didodecyl-9H-fluorene

A169408-100G A169408-10G A169408-1G A169408-25G A169408-5G
77028-946EA 132.62 USD
77028-946 77028-948 77028-950 77028-952 77028-954
9,9-Di-n-dodecyl-2,7-dibromofluorene 97%
9,9-Di-n-dodecyl-2,7-dibromofluorene

Precaución: for Research Use Only

Formula: C₃₇H₅₆Br₂
MW: 660.65 g/mol
MDL Number: MFCD03427215
CAS Number: 286438-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR