Ethylhexyl triazone 97%

Supplier: Ambeed
Warning

Synonyms: Tris(2-ethylhexyl) 4,4',4''-(1,3,5-Triazine-2,4,6-triyltriimino)trisbenzoate, Tris(2-ethylhexyl) 4,4',4''-((1,3,5-triazine-2,4,6-triyl)tris(azanediyl))tribenzoate, 4,4',4''-(1,3,5-Triazine-2,4,6-triyltriimino)trisbenzoic acid tris(2-ethylhexyl) ester

A169223-100G A169223-5G A169223-500G A169223-25G A169223-10G
77028-936EA 79 USD
77028-936 77028-938 77600-362 77667-362 77871-848
Ethylhexyl triazone 97%
Ethylhexyl triazone

Precaución: for Research Use Only

Formula: C₄₈H₆₆N₆O₆
MW: 823.09 g/mol
MDL Number: MFCD09753106
CAS Number: 88122-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR