Methyl-2-(2-(((benzyloxy)carbonyl)amino)propan-2-yl)-5-hydroxy-6-oxo-1,6-dihydropyrimidine-4-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(2-(((benzyloxy)carbonyl)amino)propan-2-yl)-5-hydroxy-6-oxo-1,6-dihydropyrimidine-4-carboxylate

A169220-1G A169220-5G
77028-932EA 11.67 USD
77028-932 77028-934
Methyl-2-(2-(((benzyloxy)carbonyl)amino)propan-2-yl)-5-hydroxy-6-oxo-1,6-dihydropyrimidine-4-carboxylate 97%
Methyl-2-(2-(((benzyloxy)carbonyl)amino)propan-2-yl)-5-hydroxy-6-oxo-1,6-dihydropyrimidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₁₉N₃O₆
MW: 361.3493 g/mol
MDL Number: MFCD12026362
CAS Number: 519032-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR