Nɑ-Benzoyl-L-arginine ethyl ester hydrochloride 98%

Supplier: Ambeed
Warning

A168972-100G A168972-25G A168972-5G A168972-1G A168972-10G
76998-156EA 161 USD
76998-156 76998-158 76998-160 77600-358 77671-386
Nɑ-Benzoyl-L-arginine ethyl ester hydrochloride 98%
Nalpha-Benzoyl-L-arginine ethyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₅H₂₂N₄O₃
MDL Number: MFCD00012579
CAS Number: 2645-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR