Methyl 2-amino-2,3-dihydro-1H-indene-2-carboxylate hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-indan-2-carboxylic acid methyl ester hydrochloride

A168834-100G A168834-100MG A168834-1G A168834-250MG A168834-25G A168834-5G
76985-324EA 1289 USD
76985-324 76985-326 76985-328 76985-330 76985-332 76985-334
Methyl 2-amino-2,3-dihydro-1H-indene-2-carboxylate hydrochloride 95%
Methyl 2-amino-2,3-dihydro-1H-indene-2-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₄ClNO₂
MW: 227.68 g/mol
MDL Number: MFCD08437619
CAS Number: 199330-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR