4-Aminopyrimidine-5-carboxylic acid 95%

Supplier: Ambeed
Warning

A168632-100MG A168632-10G A168632-250MG A168632-5G
77011-530EA 8.49 USD
77011-530 77011-532 77011-534 77011-536
4-Aminopyrimidine-5-carboxylic acid 95%
4-Aminopyrimidine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅N₃O₂
MDL Number: MFCD00047383
CAS Number: 20737-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR