2-Fluoro-5-nitro-6-picoline 98%

Supplier: Ambeed
Warning

Synonyms: 6-Fluoro-2-methyl-3-nitropyridine, 2-Fluoro-6-methyl-5-nitropyridine

A168563-100G A168563-10G A168563-1G A168563-250MG A168563-25G A168563-5G
76998-138EA 280 USD
76998-138 76998-140 76998-142 76998-144 76998-146 76998-148
2-Fluoro-5-nitro-6-picoline 98%
2-Fluoro-5-nitro-6-picoline

Precaución: for Research Use Only

Formula: C₆H₅FN₂O₂
MW: 156.12
MDL Number: MFCD03095071
CAS Number: 18605-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR