2-Chloro-3-ethoxy-6-fluorophenylboronic acid 97%

Supplier: Ambeed
Warning

A168548-1G A168548-250MG A168548-5G
77028-924EA 42.44 USD
77028-924 77028-926 77028-928
2-Chloro-3-ethoxy-6-fluorophenylboronic acid 97%
2-Chloro-3-ethoxy-6-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BClFO₃
MW: 218.4200 g/mol
MDL Number: MFCD09800880
CAS Number: 957120-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR