3-Chloro-2-fluorobenzotrifluoride 97%

Supplier: Ambeed
Warning

Synonyms: 1-Chloro-2-fluoro-3-(trifluoromethyl)benzene

A168496-100MG A168496-1G A168496-250MG A168496-5G
76998-128EA 23.34 USD
76998-128 76998-130 76998-132 76998-134
3-Chloro-2-fluorobenzotrifluoride 97%
3-Chloro-2-fluorobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃ClF₄
MW: 198.5500 g/mol
MDL Number: MFCD11100532
CAS Number: 1099597-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR