(R)-1-(3-(Trifluoromethyl)phenyl)ethanamine hydrochloride 95%

Supplier: Ambeed
Warning

A168419-100MG A168419-1G A168419-250MG A168419-5G
76985-312EA 66.84 USD
76985-312 76985-314 76985-316 76985-318
(R)-1-(3-(Trifluoromethyl)phenyl)ethanamine hydrochloride 95%
(R)-1-(3-(Trifluoromethyl)phenyl)ethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₁ClF₃N
MW: 225.63 g/mol
MDL Number: MFCD11100947
CAS Number: 1213630-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR