1-Boc-4-(methylamino)piperidine 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Methylamino)piperidine, N1-BOC protected, tert-Butyl 4-(methylamino)piperidine-1-carboxylate

A168393-100G A168393-10G A168393-1G A168393-25G A168393-5G
76998-118EA 159.14 USD
76998-118 76998-120 76998-122 76998-124 76998-126
1-Boc-4-(methylamino)piperidine 98%
1-Boc-4-(methylamino)piperidine

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.3046 g/mol
MDL Number: MFCD02259411
CAS Number: 147539-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR