tert-butyl N-ethenylcarbamate 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl vinylcarbamate

A168221-1G A168221-250MG A168221-5G A168221-25G
77028-902EA 18 USD
77028-902 77028-904 77028-906 77567-460
tert-butyl N-ethenylcarbamate 98%
tert-butyl N-ethenylcarbamate

Precaución: For research use only.

Formula: C₇H₁₃NO₂
MW: 143.184 g/mol
MDL Number: MFCD00081075
CAS Number: 7150-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR