Benzyl (3S)-3-hydroxypiperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: (S)-1-Cbz-3-hydroxypiperidine

A168132-1G A168132-25G A168132-5G
77011-520EA 19 USD
77011-520 77011-522 77011-524
Benzyl (3S)-3-hydroxypiperidine-1-carboxylate 95%
Benzyl (3S)-3-hydroxypiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₃
MW: 235.279 g/mol
MDL Number: MFCD11112281
CAS Number: 94944-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR