4-Hydroxy-3-methoxy-2-nitrobenzaldehyde 95%

Supplier: Ambeed
Warning

A168106-1G A168106-250MG A168106-5G A168106-100MG A168106-25G
76985-302EA 12 USD
76985-302 76985-304 76985-306 77885-328 77885-329
4-Hydroxy-3-methoxy-2-nitrobenzaldehyde 95%
4-Hydroxy-3-methoxy-2-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇NO₅
MDL Number: MFCD00017020
CAS Number: 2450-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR