L-isoleucine benzyl ester tosylate 95%

Supplier: Ambeed
Warning

Synonyms: (2S,3S)-Benzyl 2-amino-3-methylpentanoate 4-methylbenzenesulphonate, H-Ile-OBzl·p-tosylate, H-lle-OBzl·Tos

A167973-100G A167973-1G A167973-25G
76998-092EA 186 USD
76998-092 76998-094 76998-096
L-isoleucine benzyl ester tosylate 95%
L-isoleucine benzyl ester tosylate

Precaución: for Research Use Only

Formula: C₂₀H₂₇NO₅S
MW: 393.498 g/mol
MDL Number: MFCD00050511
CAS Number: 16652-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR