1-Chloro-2-(chloromethyl)-3-nitrobenzene 95%

Supplier: Ambeed
Danger

A167972-100MG A167972-1G
77011-516EA 44 USD
77011-516 77011-518
1-Chloro-2-(chloromethyl)-3-nitrobenzene 95%
1-Chloro-2-(chloromethyl)-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₂
MDL Number: MFCD00190097
CAS Number: 15258-72-7
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR