Ethyl 2-aminobenzo[d]thiazole-7-carboxylate 98%

Supplier: Ambeed
Warning

A167902-100MG A167902-1G
77011-508EA 13 USD
77011-508 77011-510
Ethyl 2-aminobenzo[d]thiazole-7-carboxylate 98%
Ethyl 2-aminobenzo[d]thiazole-7-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂S
MDL Number: MFCD13195422
CAS Number: 677304-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR