5-Fluoro-2-methylisonicotinic acid 97%

Supplier: Ambeed
Warning

A167849-1G A167849-250MG A167849-100MG
77011-504EA 845.54 USD
77011-504 77011-506 77683-376
5-Fluoro-2-methylisonicotinic acid 97%
5-Fluoro-2-methylisonicotinic acid

Precaución: For research use only.

Formula: C₇H₆FNO₂
MW: 155.126 g/mol
MDL Number: MFCD18256823
CAS Number: 885588-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR