2-Bromo-5-iodo-4-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-5-iodo-4-picoline

A167795-100MG A167795-1G A167795-250MG A167795-5G
77011-496EA 8.49 USD
77011-496 77011-498 77011-500 77011-502
2-Bromo-5-iodo-4-methylpyridine 98%
2-Bromo-5-iodo-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅BrIN
MW: 297.92 g/mol
MDL Number: MFCD09037465
CAS Number: 942206-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR