5-Chloro-6-methoxypyridine-3-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A167784-1G A167784-250MG A167784-5G
77028-888EA 87 USD
77028-888 77028-890 77028-892
5-Chloro-6-methoxypyridine-3-boronic acid pinacol ester 98%
5-Chloro-6-methoxypyridine-3-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BClNO₃
MDL Number: MFCD08063054
CAS Number: 1083168-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR