6-Bromoisoquinolin-5-amine 95%

Supplier: Ambeed
Warning

A167677-100MG A167677-250MG
76985-290EA 38 USD
76985-290 76985-292
6-Bromoisoquinolin-5-amine 95%
6-Bromoisoquinolin-5-amine

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MDL Number: MFCD14706163
CAS Number: 850198-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR