Methyl 6-fluoro-1H-indazole-3-carboxylate 98%

Supplier: Ambeed
Warning

A167655-100MG A167655-1G A167655-250MG
77011-490EA 23.34 USD
77011-490 77011-492 77011-494
Methyl 6-fluoro-1H-indazole-3-carboxylate 98%
Methyl 6-fluoro-1H-indazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇FN₂O₂
MW: 194.162 g/mol
MDL Number: MFCD07371604
CAS Number: 885279-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR