(3-(Benzyloxy)phenyl)methanamine hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A167646-1G
A167646-250MG
76985-286EA
144
USD
76985-286
76985-288
(3-(Benzyloxy)phenyl)methanamine hydrochloride 95%
(3-(Benzyloxy)phenyl)methanamine hydrochloride
Precaución: for Research Use Only
Formula:
C₁₄H₁₆ClNO |
MDL Number:
MFCD15732122 CAS Number: 104566-41-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...