2-(2-(4-Benzhydrylpiperazin-1-yl)ethoxy)ethanol dihydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Decloxizine dihydrochloride
A167612-100MG
A167612-1MG
A167612-50MG
76985-280EA
91.24
USD
76985-280
76985-282
76985-284
2-(2-(4-Benzhydrylpiperazin-1-yl)ethoxy)ethanol dihydrochloride 98%
2-(2-(4-Benzhydrylpiperazin-1-yl)ethoxy)ethanol dihydrochloride
Precaución: for Research Use Only
Formula:
C₂₁H₃₀Cl₂N₂O₂ MW: 413.38 g/mol |
MDL Number:
MFCD02183705 CAS Number: 13073-96-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...