2-(4-aminophenoxy)-2-methylpropanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Aminophenoxy)-2-methylpropanoic acid

A167608-100MG A167608-1G A167608-250MG
77011-480EA 18.04 USD
77011-480 77011-482 77011-484
2-(4-aminophenoxy)-2-methylpropanoic acid 95%
2-(4-aminophenoxy)-2-methylpropanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃
MW: 195.215 g/mol
MDL Number: MFCD00084883
CAS Number: 117011-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR