Boc-(S)-2-amino-3-methoxypropionicacid dicyclohexylammonium salt 95%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(Boc-amino)-3-methoxypropionic acid·DCHA, Boc-Ser(Me)-OH·DCHA, Boc-2-amino-3-methoxypropionicacid-dicyclohexylammonium-salt, Boc-O-methyl-L-S-dicyclohexylammonium-salt, Boc-O-methyl-L-serine (dicyclohexylammonium) salt, N-tert-Butoxycarbonyl-O-methyl-L-serine·DCHA, Boc-2-amino-3-methoxypropionic acid·DCHA, Boc-O-methyl-L-serine·DCHA, Dicyclohexylamine (S)-2-((tert-butoxycarbonyl)amino)-3-methoxypropanoate

A167447-1G A167447-5G
76985-270EA 6.37 USD
76985-270 76985-272
Boc-(S)-2-amino-3-methoxypropionicacid dicyclohexylammonium salt 95%
Boc-(S)-2-amino-3-methoxypropionicacid dicyclohexylammonium salt

Precaución: for Research Use Only

Formula: C₉H₁₇NO₅·C₁₂H₂₃N
MW: 400.56 g/mol
MDL Number: MFCD09260875
CAS Number: 69912-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR