(±)-3-Fluoromandelic acid 95%

Supplier: Ambeed
Warning

Synonyms: (±)-3-Fluorophenylglycolic acid, 2-(3-Fluorophenyl)-2-hydroxyacetic acid

A167099-100MG A167099-10G A167099-1G A167099-250MG A167099-5G
76998-058EA 5 USD
76998-058 76998-060 76998-062 76998-064 76998-066
(±)-3-Fluoromandelic acid 95%
(±)-3-Fluoromandelic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₃
MDL Number: MFCD03095383
CAS Number: 395-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR