1-(2-Fluoroethyl)piperazine dihydrochloride 95%

Supplier: Ambeed
Warning

A167033-100MG A167033-1G A167033-250MG
76985-252EA 43.5 USD
76985-252 76985-254 76985-256
1-(2-Fluoroethyl)piperazine dihydrochloride 95%
1-(2-Fluoroethyl)piperazine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃FN₂·2ClH
MW: 205.101 g/mol
MDL Number: MFCD21602355
CAS Number: 1089279-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR