5-Formyl-2-thiopheneboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-Formylthiophen-2-boronic acid, 5-Formylthiophene-2-boronic acid

A166987-100G A166987-10G A166987-250MG A166987-25G A166987-5G A166987-1G
77011-450EA 210.06 USD
77011-450 77011-452 77011-454 77011-456 77011-458 77683-367
5-Formyl-2-thiopheneboronic acid 97%
5-Formyl-2-thiopheneboronic acid

Precaución: For research use only.

Formula: C₅H₅BO₃S
MW: 155.97 g/mol
MDL Number: MFCD02093666
CAS Number: 4347-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR