(3,5-Difluoro-4-nitrophenyl)methanol 97%

Supplier: Ambeed
Warning

A166916-1G A166916-250MG A166916-5G
77011-444EA 59 USD
77011-444 77011-446 77011-448
(3,5-Difluoro-4-nitrophenyl)methanol 97%
(3,5-Difluoro-4-nitrophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₃
MDL Number: MFCD18633079
CAS Number: 1123172-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR