1,7-Dichloroisoquinoline 95%

Supplier: Ambeed
Warning

A166777-100MG A166777-1G A166777-250MG A166777-5G
77028-866EA 10 USD
77028-866 77028-868 77028-870 77028-872
1,7-Dichloroisoquinoline 95%
1,7-Dichloroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₅Cl₂N
MW: 198.049 g/mol
MDL Number: MFCD11846494
CAS Number: 70810-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR